Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Enhance the operation of Refrigerated and Heated Bath Circulators with accessories that include bridges, platforms, cooling coils and connectivity solutions.
Supplier:  Thermo Scientific™ 1600018
 View more versions of this product
Catalog No. 13265124
Only null left
Description
Description
Enhance the operation of Refrigerated and Heated Bath Circulators with Thermo Scientific™ Accessories which include bridges, platforms, cooling coils and connectivity solutions.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
SC, AC, PC immersion Circulators
Adjustable Bridge
300 to 800 mm
Product Suggestions
Product Suggestions