Learn More
Digi-Sense™ Accessories for the 20250-40 Digi-Sense™ Temperature Humidity Touch-screen Recorder
Catalog No. 13245302
Change view
Click to view available options
Product Type:
Extension Sensor Cable
Replacement USB Cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
13245302 Extension Sensor Cable
13245305 Replacement USB Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13245302 Supplier Digi-Sense™ Supplier No. 2025047
The touch-screen reader has 8GB on board memory to provide virtually unlimited data logging capacity
  • Includes A2LA accredited NIST-Traceable Calibration Certificate
  • Measures, logs, and displays temperature, humidity, and dew point
  • Removable sensor for local or remote measurements using extension cable included
  • Wall mount or desk mount operation
  • Resident self-booting software, no installation necessary
  • Comes with NIST calibration
  • Stored data downloads to computer via USB cable provided in CSV format
Warranty
2 Years
Specifications
For Use With (Application) Extension Cable for Temperature/RH Touch Screen Recorder with NIST Traceable Calibration
Product Type Extension Sensor Cable
For Use With (Equipment) Temperature/RH Touch Screen Recorder with NIST Traceable Calibration (20250-40)