Learn More
Conair™ Accessories and Replacements for Waring™ Blenders::Stainless-steel Container with Ground Path
Catalog No. 1450937
Change view
Click to view available options
For Use With (Equipment):
Waring™ blender
Catalog No. For Use With (Equipment)
1450937 Waring™ blender
1 options
Catalog No. 1450937 Supplier Conair™ Supplier No. HL515
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Hazardous Location Blender Motor to comply with UL listing for Class 1, Group D hazardous locations. Stainless steel. Cloverleaf design. Capacity, 1L (32 oz.).
Specifications
Product Type Container
Capacity (Metric) 1000 mL
Material Stainless Steel
For Use With (Equipment) Waring™ blender