Learn More
Metrohm Accessories and Replacement Parts for Models 756 and 831 KF Coulometers
Catalog No. BR20928972
Change view
Click to view available options
Type:
Electrode, With Diaphragm
Electrode, Without Diaphragm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
BR20928972 Electrode, Without Diaphragm
BR20928964 Electrode, With Diaphragm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BR20928972 Supplier Metrohm Supplier No. 6.0345.100
Specifications
Type Electrode, Without Diaphragm
Temperature (English) Range -4°F to 158°F
Temperature (Metric) Range -20°C to +70°C
For Use With (Equipment) 756 and 831 Coulometric Karl Fischer titrator