Learn More
Labconco™ Accessories for Labconco™ Fiber Glass Desiccator
Catalog No. 16307243
Change view
Click to view available options
Product Type:
Desiccator Vacuum Tubing Kit
Stacking Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16307243 Desiccator Vacuum Tubing Kit
16307242 Stacking Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16307243 Supplier Labconco™ Supplier No. 5535700
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vacuum Tubing Kit allows connection of Vacuum Desiccator's valve to an accessory pump.
Vacuum Tubing Kit includes 1/4 in. ID and 1/2 in. ID vacuum tubing, each 48 in. long, two each 1/4 in. and 1/2 in. tubing clamps; 1/4 in. to 1/2 in. barbed reducer. Kit allows connection of Labconco Vacuum Desiccator's 1/4 in. diameter brass needle valve to an accessory pump with 1/2 in. connector.
Order Info
Dimensions: 12L x 12 in. W
Specifications
Product Type Desiccator Vacuum Tubing Kit
For Use With (Equipment) Labconco Fiber Glass Desiccator
Includes 0.25 in. I.D. and 0.5 in. I.D. vacuum tubing, each 48 in. long, two each 0.25 in. and 0.5 Tubing clamps, 0.25 to 0.5 in. barbed reducer
Weight (English) 5.95 lb.
Weight (Metric) 2.7 kg