Learn More
Labconco™ Accessories for Labconco™ Fiber Glass Desiccator
Catalog No. 16307242
Change view
Click to view available options
Product Type:
Desiccator Vacuum Tubing Kit
Stacking Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16307242 Stacking Rack
16307243 Desiccator Vacuum Tubing Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16307242 Supplier Labconco™ Supplier No. 5530600
Vacuum Tubing Kit allows connection of Vacuum Desiccator's valve to an accessory pump.
Powder-coated steel rack allows one Labconco Vacuum Desiccator to be placed on top of another.
Order Info
Dimensions: 12L x 12 in. W
Specifications
Product Type Stacking Rack
Material Epoxy-coated Steel
For Use With (Equipment) Labconco Fiber Glass Desiccator
Weight (English) 5.07 lb.
Weight (Metric) 2.3 kg