missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Isotemp™ Gas Tank Switcher
Description
- Napco 8000; Water Jacket Models Only
Specifications
Specifications
| For Use With (Equipment) | Napco 8000; Water Jacket Models Only |
| Type | Gas Tank Switcher |
| Electrical Requirements | 120 V, 60 Hz |
| Voltage | 120 V |
| Frequency | 60 Hz |