missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Accessories for Fisherbrand™ Multi Platform Shaker

Catalog No. 88861151
Change view
Click to view available options
Product Type:
Adjustable Tube Rack
Fixed Tube Rack
Flask Clamp
Funnel Clamp
Infusion Bottle Clamp
Platform with Adjustable Bars
Platform with Clamps
Platform with Rubber Mat
Holds:
1000 mL Flask
1000 mL Infustion Bottle
12 x 100 mL Flasks, 150 mL Beakers
2000 mL Flask
3000 mL Flask
40 x 14 mm dia. Tubes
40 x 16 mm dia. Tubes
500 mL Flask
500 mL Infusion Bottle
6 x 250 mL Flasks, 150 mL Beakers
Funnels
Vessels
5000 mL
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Holds
88861151 Adjustable Tube Rack 40 x 16 mm dia. Tubes
88861131 Platform with Rubber Mat Vessels
88861132 Platform with Clamps 6 x 250 mL Flasks, 150 mL Beakers
88861133 Platform with Clamps 12 x 100 mL Flasks, 150 mL Beakers
88861134 Platform with Adjustable Bars Vessels
88861139 Flask Clamp 500 mL Flask
88861140 Flask Clamp 1000 mL Flask
88861141 Flask Clamp 2000 mL Flask
88861142 Flask Clamp 3000 mL Flask
88861143 Flask Clamp 5000 mL
88861145 Infusion Bottle Clamp 500 mL Infusion Bottle
88861146 Infusion Bottle Clamp 1000 mL Infustion Bottle
88861147 Funnel Clamp Funnels
88861148 Fixed Tube Rack 40 x 14 mm dia. Tubes
88861149 Fixed Tube Rack 40 x 16 mm dia. Tubes
88861150 Adjustable Tube Rack 40 x 14 mm dia. Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 88861151 Supplier Fisherbrand™ Supplier No. 88861151
Get an Accessory with Shaker Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accessories required for the use of Fisher Scientific™ Multi Platform Shaker

Accessories for multi platform shaker to enhance performance and flexibility

  • Increase performance and flexibility

Specifications

Holds 40 x 16 mm dia. Tubes
Product Type Adjustable Tube Rack