missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Accessories for Combisart™ Systems: Filtrate Flasks

Catalog No. 14555899
Change view
Click to view available options
Capacity (Metric):
1000mL
2000mL
5000mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14555899 5000mL
14555892 1000mL
14555898 2000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14555899 Supplier Sartorius Supplier No. 166721

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Filtrate flasks for use with Sartorius Combisart Systems

  • Suction flask for individual filter holders
  • Vacuum-resistant borosilicate glass, 3.3 (Duran 50 glass)
  • Plastic hose nipple acc. to DIN 12476
  • Stopper/tube
  • Sterilizable

Specifications

Product Type Suction Flask
Display Type None
Capacity (Metric) 5000mL
Color Translucent
Material Borosilicate Glass
For Use With (Application) For collecting the filtrate
For Use With (Equipment) Individual vacuum filter holder
Includes Plastic Hose Nipple