missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Accessories for Combisart™ Systems: Filtrate Flasks
Filtrate flasks for use with Sartorius Combisart Systems
Supplier: Sartorius 16672
Description
Filtrate flasks for use with Sartorius Combisart Systems
- Suction flask for individual filter holders
- Vacuum-resistant borosilicate glass, 3.3 (Duran 50 glass)
- Plastic hose nipple acc. to DIN 12476
- Stopper/tube
- Sterilizable
Chemical Identifiers
| Suction Flask | |
| Translucent | |
| For collecting the filtrate |
| None | |
| Borosilicate Glass | |
| Individual vacuum filter holder |
Specifications
| Suction Flask | |
| 2000mL | |
| Borosilicate Glass | |
| Individual vacuum filter holder |
| None | |
| Translucent | |
| For collecting the filtrate | |
| 9 mm Plastic Safety Hose Nipple |