Learn More
Thermo Scientific™ Accessories for Cimarec+™ Stirrer Series
Get precise stirring control for your routine protocols.
Supplier:  Thermo Scientific™ CIC0001445
 View more versions of this product
Catalog No. CIC0001445
Only null left
Description
Description
These accessories are for use with the Cimarec+™ series digital stirrers. Our new Thermo Scientific™ Cimarec+™ series digital stirrers offer precise stirring control for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operation.
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strongmagnetic coupling
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for stirring
  • Raised display design protects electronics from spills
  • Optional splash guard protection shield
  • Variable sizes for different sample volumes
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Chemical Identifiers
LED
Specifications
LED
Transparent shield for 7x7
Product Suggestions
Product Suggestions