missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Access Port
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KB65, KB130, KB260, KB470, KB720, and KB 1060
Specifications
Specifications
| For Use With (Equipment) | KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060 |
| Material | Silicone |
| Type | Access Port |
| Ports | 1 on Left Side |
| Port Size | 50 mm |