missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Access Port

Catalog No. 502875758
Change view
Click to view available options
Port Size:
30 mm
50 mm
100 mm
Ports:
1 on Left Side
1 on Right Side
1 on Top
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Port Size Ports
502875758 50 mm 1 on Right Side
502875587 30 mm 1 on Left Side
502875695 50 mm 1 on Left Side
502875669 100 mm 1 on Left Side
502875651 30 mm 1 on Right Side
502875606 100 mm 1 on Right Side
502875668 50 mm 1 on Top
502875685 100 mm 1 on Top
502875653 30 mm 1 on Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 502875758 Supplier BINDER™ Supplier No. 80122485

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Access port with silicone plug for use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600.

  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600

Specifications

For Use With (Equipment) KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Material Silicone
Type Access Port
Ports 1 on Right Side
Port Size 50 mm