missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Access Port

Catalog No. 502875736
Change view
Click to view available options
Port Size:
30 mm
50 mm
100 mm
Ports:
1 on Left Side
1 on Right Side
1 on Top
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Port Size Ports
502875736 100 mm 1 on Top
502875619 30 mm 1 on Left Side
502875589 50 mm 1 on Left Side
502875670 100 mm 1 on Left Side
502875588 30 mm 1 on Right Side
502875735 50 mm 1 on Right Side
502875652 100 mm 1 on Right Side
502875684 50 mm 1 on Top
502875607 30 mm 1 on Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 502875736 Supplier BINDER™ Supplier No. 80122496

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Access port with silicone plug for use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600.

  • Round access port with silicone plug
  • Choose between 30, 50, or 100 mm plug
  • Available on left, right, or top of the chamber
  • For use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600

Specifications

For Use With (Equipment) KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600
Material Silicone
Type Access Port
Ports 1 on Top
Port Size 100 mm