Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Catalog No. ACE34567
Change view
Click to view available options
For Use With (Equipment):
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series
TSX Universal
Type:
Access Key Pack (EU) Standalone Cards
Access Key Pack (U.S.) Standalone Cards
Factory installed. No stand-alone orders.
Factory-Installed. No stand-alone orders
Field-installed
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
ACE34567 TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series Access Key Pack (EU) Standalone Cards
ACU34567 TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series Access Key Pack (U.S.) Standalone Cards
RAC34567 TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series Factory installed. No stand-alone orders.
ELTSXU TSX Universal Factory-Installed. No stand-alone orders
FELTSXU TSX Universal Field-installed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ACE34567 Supplier Thermo Scientific™ Supplier No. ACE34567
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
  • Card access comes with 5 key cards
  • Field installed - purchased separately
  • Factory installed in the plant with purchase of unit
Specifications
Type Access Key Pack (EU) Standalone Cards
For Use With (Equipment) TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series
Includes Includes 5 cards, supports ISO14443 Protocol