Learn More
United Scientific™ Acceleration Line Trolley
Catalog No. S000865
Change view
Click to view available options
Dimensions (L x W x H):
6 x 3.5 x 1 in.
Catalog No. Dimensions (L x W x H)
S000865 6 x 3.5 x 1 in.
1 options
Prop 65
Catalog No. S000865 Supplier United Scientific™ Supplier No. ACCTRLLY
Prop 65
The acceleration trolley rides down a long 30-foot cable with steel spring clamps on the ends.
The angle of an incline has a direct effect on the acceleration due to gravity of a freely moving object. When the ends of the cables are attached to a secure position, this investigates how the acceleration down the cable varies based on the angle of the cable. Vary the mass of the trolley by using bottom holes to attach hanging weights (required but not included). Weights of 100 g up to 250 g yield the best results. An index card can be attached to the top bar and utilized as a photogate to make more accurate time measurements. The overall dimensions of the trolley are 6 x 3.5 x 1 in.
  • Spring clips make connections easy
  • Hanging masses can be attached to the built-in holes
  • Includes activity guide
Specifications
Product Type Acceleration Line Trolley
Material Steel
Dimensions (L x W x H) 6 x 3.5 x 1 in.