missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acarbose, 95%

Catalog No. AAJ6173703
Change view
Click to view available options
Quantity:
1 g
2 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6173703 1 g
AAJ6173704 2 g
AAJ6173706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6173703 Supplier Thermo Scientific Chemicals Supplier No. J6173703

A reversible alpha-glucosidase inhibitor | CAS: 56180-94-0 | C25H43NO18 | 645.608 g/mol

A reversible alpha-glucosidase inhibitorAcarbose is used as a reversible alfa-glucosidase inhibitor. It acts as an anti-diabetic drug, which finds useful application to treat type 2 diabetes mellitus and prediabetes. It is also employed for Hepatitis. It also acts as a beta glucosidase inhibitor, antihyperlipidaemia and antiobesity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reversible alpha-glucosidase inhibitorAcarbose is used as a reversible alfa-glucosidase inhibitor. It acts as an anti-diabetic drug, which finds useful application to treat type 2 diabetes mellitus and prediabetes. It is also employed for Hepatitis. It also acts as a beta glucosidase inhibitor, antihyperlipidaemia and antiobesity.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 56180-94-0
Molecular Formula C25H43NO18
Molecular Weight (g/mol) 645.608
MDL Number MFCD00869592
InChI Key XUFXOAAUWZOOIT-AKWDJIIVSA-N
Synonym acarbose
PubChem CID 131674886
IUPAC Name (3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
SMILES CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO

Specifications

Color White
Quantity 1 g
Merck Index 14,18
Solubility Information Soluble in water.
Formula Weight 645.61
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Acarbose
EINECSNumber 260-030-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only