missing translation for 'onlineSavingsMsg'
Learn More

Acarbose, 95%

Catalog No. p-7077774
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459080010 1 g
AC459080050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459080010 Supplier Thermo Scientific Chemicals Supplier No. 459080010

CAS: 56180-94-0 | C25H43NO18 | 645.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56180-94-0
Molecular Formula C25H43NO18
Molecular Weight (g/mol) 645.61
InChI Key XUFXOAAUWZOOIT-AKWDJIIVSA-N
Synonym acarbose
PubChem CID 131674886
IUPAC Name (3S,5S)-5-[(2R,3S,5S)-5-[(2R,3S,5S)-3,4-dihydroxy-6-methyl-5-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]oxan-2-yl]oxy-3,4-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
SMILES CC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)NC4C=C(C(C(C4O)O)O)CO

Specifications

Melting Point 90.0°C
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation +168° to +183° (20°C, 589nm) (1% in water)
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in ethanol
Formula Weight 645.61
Percent Purity 95%
Chemical Name or Material Acarbose
Show More Show Less
EINECSNumber 260-030-7

RUO – Research Use Only