missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ AC800 ESD Nitrile-Coated Cut Resistant Gloves

Catalog No. p-200106360
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18139742 Small
18139741 Medium
18139740 Large
18139743 X-Large
18139744 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18139742 Supplier SHOWA™ Supplier No. AC800L08
Only null left

Electrostatic dissipative 18-gauge HPPE, cut resistant embossed microporous foam nitrile coating glove

The new AC800 is an 18-gauge seamless knit glove made using HPPE blended with static dissipative yarn and featuring an embossed microporous foam nitrile palm dip. The unique features of these elements allow for this product to be touchscreen friendly, silicone free, fingerprint free and contains no natural rubber latex. Certified under the standard EN 16350, the AC800 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Nitrile Foam
Color Gray
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material HPPE
Powdered Powder Free
Product Type ESD Gloves
Size Small
Sterility Non-sterile
Thickness (English) 18 gauge
Thickness (Metric) 0.8 mm
Show More Show Less