Learn More
AirClean™ Systems AC600 Series Vertical Laminar Flow Fume Hood
Catalog No. 361004377
Change view
Click to view available options
Dimensions (D x W x H):
24 x 32 x 30 in.
24 x 32 x 39 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
361004377 24 x 32 x 30 in.
361004378 24 x 32 x 39 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361004377 Supplier AirClean™ Systems Supplier No. AC632LFC
For applications that are not hazardous to operator but require clean work area, choose an Airclean AC600 Series Vertical Laminar Flow Fume Hood.
  • UVTect™ microprocessor controller
  • Class 100 (ISO 5) vertical laminar flow
  • 360-degree visibility
  • Integral polypropylene base for easy cleaning
  • Supplied fully assembled
  • Built-in fluorescent light
  • HEPA filter monitor automatically indicates when filter change is required
  • Available in 110V or 220V versions
  • Compact, light weight design allows for multiple “clean zones” within laboratory
Options
  • Shortwave UV light
  • Custom sturdy cart allows unit to be made portable; Cart sold separately, see 36-100-0054
  • Polypropylene - metal free construction
  • ULPA filter
  • Sliphatch access port
Certifications
UL listed, CSA, CE marked
Compliance
Certified to meet or exceed applicable clean bench standards.
Specifications
Material Thermoplastic
Certifications/Compliance ISO 5 and FS209E, USP 797
Dimensions (D x W x H) 24 x 32 x 30 in.
Voltage 110/220 V