missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ AC200 ESD Uncoated Cut Resistant Gloves

Catalog No. 18139738
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18139738 Small
17018188 Medium
17018187 Large
18139739 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18139738 Supplier SHOWA™ Supplier No. AC200S06
Only null left

Electrostatic dissipative 18-gauge HPPE, uncoated cut resistant glove

The new SHOWA AC200 has an 18-gauge seamless knit, cut-resistant liner with UHMWPE static dissipative yarn. It is free of natural rubber latex and is touchscreen compatible, silicone-free and fingerprint-free. Certified under the standard EN 16350, the AC200 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color Gray
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material HPPE
Powdered Powder Free
Product Type ESD Gloves
Size Small
Sterility Non-sterile
Thickness (English) 18 gauge
Thickness (Metric) 0.8 mm
Show More Show Less