Learn More
SHOWA™ AC200 ESD Uncoated Cut Resistant Gloves
Electrostatic dissipative 18-gauge HPPE, uncoated cut resistant glove
Supplier:  SHOWA™ AC200L08
 View more versions of this product
Catalog No. 17-018-187
Only null left
Explore available promotions
Description
Description
The new SHOWA AC200 has an 18-gauge seamless knit, cut-resistant liner with UHMWPE static dissipative yarn. It is free of natural rubber latex and is touchscreen compatible, silicone-free and fingerprint-free. Certified under the standard EN 16350, the AC200 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).
Specifications
Specifications
Yes
Uncoated
Reusable
Ambidextrous
HPPE
ESD Gloves
Non-sterile
0.8 mm
Industrial
Gray
No Texture
Yes
Powder Free
Large
18 gauge
Product Suggestions
Product Suggestions