missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ AC/DC Generator Demonstration Set

Catalog No. S65999
Change view
Click to view available options
Includes:
Activity guide, clear base
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S65999 Activity guide, clear base
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65999 Supplier United Scientific™ Supplier No. ACDCSET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An effective hand generator for AC/DC demonstrations.

Simple yet effective, this hand generator offers a unique way to teach how AC and DC electricity is produced.

  • Clear base for easy demonstration of connections
  • Banana plugs can be inserted into different connectors to change from AC to DC electricity
  • Includes an activity guide
  • Dimensions: 10 x 7 x 9 in. (25 x 17.5 x 22.5cm)

Specifications

For Use With (Application) Demonstrates how AC and DC electricity is produced
Includes Activity guide, clear base
Product Type ACDC Demonstrator