missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorbent Surface Liners

Catalog No. 14127501
Change view
Click to view available options
Dimensions (L x W):
300 ft. x 20 in.
24 x 20 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
14127501 300 ft. x 20 in.
14127500 24 x 20 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14127501 Supplier Fisherbrand™ Supplier No. AL20600
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Economical, highly absorbent bench liners protect surfaces from spills, allowing for safe and easy cleanup.

  • Soft, durable, and highly absorbent multilayer liner cushions surfaces and absorbs and retains spills
  • Waterproof polypropylene backing resists most chemicals
  • Easily cut to desired size
  • White
  • Ideal for protecting benches and surfaces
  • Available in roll or dispenser box

Specifications

Product Type Bench Liner
Material Cellulose/Poly Backing
Absorbancy 380 mL/m2
Length (English) 300 ft.
Dimensions (L x W) 300 ft. x 20 in.
Color White
For Use With (Application) Protects benchtops, floors and carts with this highly absorbent cover
Quantity 1 Roll