Learn More
Alkali Scientific ASI™ Absorbent Bench-Top Liner, 20"" Wide, 50' Roll in Dispenser Box, 6/Case
Catalog No. 503026504
Encompass
Protect surfaces from spills with economical, highly absorbent. bench liners that allow for safe, easy cleanup Soft, durable, and highly absorbent multilayer liner cushions surfaces and absorbs and retains spills Waterproof polypropylene backing resists most chemicals Easily cut to desired size White Ideal for protecting benches and surfaces Available in roll or dispenser box
Catalog No. 503026504 Supplier Alkali Scientific Supplier No. ABL50D6
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Protect surfaces from spills with economical, highly absorbent. bench liners that allow for safe, easy cleanup Soft, durable, and highly absorbent multilayer liner cushions surfaces and absorbs and retains spills Waterproof polypropylene backing resists most chemicals Easily cut to desired size White Ideal for protecting benches and surfaces Available in roll or dispenser box