Learn More
Alkali Scientific ASI™ Absorbent Bench-Top Liner, 20"" Wide, 300' Roll, 1/Pack
Catalog No. 503026554
Encompass
Protect surfaces from spills with economical, highly absorbent. bench liners that allow for safe, easy cleanup Soft, durable, and highly absorbent multilayer liner cushions surfaces and absorbs and retains spills Waterproof polypropylene backing resists most chemicals Easily cut to desired size White Ideal for protecting benches and surfaces Available in roll or dispenser box
Catalog No. 503026554 Supplier Alkali Scientific Supplier No. ABL300
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Protect surfaces from spills with economical, highly absorbent. bench liners that allow for safe, easy cleanup Soft, durable, and highly absorbent multilayer liner cushions surfaces and absorbs and retains spills Waterproof polypropylene backing resists most chemicals Easily cut to desired size White Ideal for protecting benches and surfaces Available in roll or dispenser box