missing translation for 'onlineSavingsMsg'
Learn More

(+)-Abscisic acid, 98%

Catalog No. AC342405000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC342405000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC342405000 Supplier Thermo Scientific Chemicals Supplier No. 342405000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21293-29-8 | C15H20O4 | 264.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21293-29-8
Molecular Formula C15H20O4
Molecular Weight (g/mol) 264.32
MDL Number MFCD00066545,MFCD00066545,MFCD00075619
InChI Key JLIDBLDQVAYHNE-YKALOCIXSA-N
IUPAC Name (2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoic acid
SMILES C\C(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O

Specifications

Melting Point 160.0°C to 163.0°C
Assay Percent Range 98%
Quantity 500 mg
Merck Index 15, 12
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: soluble in aq. nahco3, chloroform, acetone, ethyl, acetate, ether, ethanol, slightly soluble in benzene., sparingly soluble in petroleum ether
Formula Weight 264.32
Chemical Name or Material (+)-Abscisic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

EINECSNumber 244-319-5

RUO – Research Use Only