missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TBARS Assay Kit
Description
Includes
- Thiobarbituric Acid 1 vial
- TBA Acetic Acid 2 vials
- TBA Sodium Hydroxide (10X) 1 vial
- TBA Malondialdehyde Standard 1 vial
- TBA SDS Solution 1 vial
- 96-Well Solid Plate (Colorimetric Assay) 1 plate
- 96-Well Solid Plate (black) 1 plate
- 96-Well Cover Sheet 2 covers
TBARS Assay Kit provides a simple, reproducible, and standardized tool for assaying lipid peroxidation in plasma, serum, urine, tissue homogenates, and cell lysates.
Applications:
Functional Study
Specifications
Specifications
| Content And Storage | 4°C |
| For Use With (Application) | Used for assaying lipid peroxidation and functional study |
| Includes | 1 Vial Thiobarbituric Acid, 2 vials TBA Acetic Acid, 1 vial TBA Sodium Hydroxide 10X, 1 vial TBA Malondialdehyde standard, 1 vial TBA SDS solution, 1 plate 96-well solid plate (colorimetric assay), 1 plate 96-Well solid plate (black), 2 covers 96-Well cover sheet |
| Quantity | 96 Assays |
| Sample Type | Plasma, Serum, Urine, Tissue Homogenates and Cell Lysates |
| Type | Detection Assay Kit |