missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Phenylalanine Assay Kit (Fluorometric)
Description
Includes
- Phenylalanine Assay Buffer 25mL
- Tyrosinase lyophilized
- Enzyme Mix lyophilized
- Developer lyophilized
- Phenyalanine Standard (10.0μmol) lyophilized
Specifications
Specifications
| Calibration Range | 2 to 300μM |
| Content And Storage | Store the kit at -20°C. |
| Description | Phenylalanine Assay Kit is a quantitative fluorimetric determination of phenylalanine at FL530/595 nm. |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | Assay Buffer (10mL), NAD Solution (1mL), Probe (750μL), Enzyme A (1 vial), Enzyme B (120μL), Standard (120μL) |
| Quantity | 1 Kit |
| Sample Type | Serum, Urine |
| Sample Volume | 10 μL |
| Type | Phenylalanine Assay Kit |