missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MTT Cell Viability Assay Kit
Description
Includes
- Assay buffer, 10mL
- Solubilizer, 50mL
- Reagent, 1 vial
Specifications
Specifications
| Content And Storage | Store Reagent at -20°C. Store Assay Buffer at 4°C. Store Solubilization Solution at room temperature. |
| Description | MTT Cell Viability Assay Kit is a non-radioactive fluorescent assay for cell proliferation and cytotoxicity. |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | 10mL assay buffer, 50mL solubilizer, reagent |
| Quantity | 1 Kit |
| Type | Cell Viability Assay Kit |