missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Invertase Assay Kit
Description
Includes
- 10x reaction buffer: (pH 4.5), 12mL
- Assay buffer, 10mL
- Glucose standard, 1mL
- 10x sucrose, 1.5mL
- Enzyme mix, 120μL
- Dye reagent ,120μL
Specifications
Specifications
| Calibration Range | 0.007U/L |
| Content And Storage | Store 10x Reaction Buffer and Assay Buffer at 4°C. Store other reagents at -20°C. |
| Description | Invertase Assay Kit is a quantitative determination of invertase/sucrase activity. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Includes | 12mL reaction buffer 6X, (pH 4.5), 10mL assay buffer, 1mL Glucose standard, 1.5mL Sucrose 6X, 120μL enzyme mix, 120μL dye reagent |
| Quantity | 1 Kit |
| Sample Type | Biological Sample |
| Sample Volume | 40 μL |
| Type | Detection Assay Kit |