missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Hemoglobin Assay Kit
Description
Includes
- Reagent, 50mL
- Calibrator, 10mL
Specifications
Specifications
| Calibration Range | 0.9 to 200mg/dL |
| Content And Storage | Store the kit at 4°C. |
| Description | Hemoglobin Assay Kit is a colorimetric determination of total hemoglobin at 400 nm. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Includes | 50mL Reagent, 10mL calibrator |
| Quantity | 1 Kit |
| Sample Type | Blood, Plasma, Serum, Urine |
| Sample Volume | 50 μL |
| Type | Detection Assay Kit |