missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Free Fatty Acid Assay Kit

For quantitative colorimetric/fluorometer fatty acid determination in cell culture, food sample, milk, plasma, saliva, serum, urine

Supplier:  Abnova™ KA1667

Catalog No. 89028399


Only null left

Includes:
  • Assay buffer, 20mL
  • Dye reagent, 120μL
  • Enzyme A, 120μL
  • Enzyme B, 120μL
  • CoSubstrate, 120μL
  • Standard, 1mL (1mM palmitic acid)

Description

Description

Specifications

Specifications

7 to 1000μM colorimetric assay, 7 to 100μM fluorimetric assay
Free Fatty Acid Assay Kit is a quantitative colorimetric/fluorimetric fatty acid determination.
Functional Study
1 Kit
10 μL
Store the kit at -20°C.
Colorimetric, Fluorometric
20mL assay buffer, 120μL dye reagent, 120μL enzyme A, 120μL enzyme B, 120μL Co substrate, 1mL 1mM Palmitic Acid standard
Cell Culture, Food Sample, Milk, Plasma, Saliva, Serum, Urine
Detection Assay Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions