missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CSF1R (Mouse) ELISA Kit

Catalog No. 89101027
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89101027 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89101027 Supplier Abnova™ Supplier No. KA3348
Only null left

Includes

  • 96-well plate precoated with anti- mouse CSF1R antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse CSF1R standard, 10ng x 2
  • Biotinylated anti- mouse CSF1R antibody(dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively mouse Csf1r. Sandwich enzyme immunoassay.

Csf1r (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Csf1r (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Csf1r.
Detection Limit 10pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernate, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Csf1r (Mouse) ELISA Kit
Specificity Natural and recombinant mouse CSF1R
No. of Assays 96
Show More Show Less