missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Creatine Kinase Assay Kit
Description
Includes
- Assay buffer, 12mL
- Substrate solution, 1mL
- Enzyme mix, 120μL
- Calibrator, 150μL
Specifications
Specifications
| Calibration Range | 5 to 300U/L |
| Content And Storage | Store the kit at -20°C. |
| Description | Creatine Kinase Assay Kit is a colorimetric determination of creatine kinase activity at 340 nm. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Includes | 12mL assay buffer, 1mL substrate solution, 120μL enzyme mix, 150μL calibrator |
| Quantity | 1 Kit |
| Sample Type | Biological Sample, Plasma, Serum |
| Type | Kinase Assay Kit |