missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ BCG Albumin Assay Kit

Catalog No. 89028344
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89028344 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89028344 Supplier Abnova™ Supplier No. KA1612

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Reagent, 50mL
  • Albumin standard, 1mL 5g/dL BSA

BCG Albumin Assay Kit is a quantitative colorimetric albumin determination at 620 nm.

  • Simple and accurate: Use as little as 5 μL samples
  • Simple and high throughput: procedure involves addition of a single working reagent and incubation for 5 minutes
  • Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day
  • Improved reagent stability and versatility: The optimized formulation has greatly enhanced reagent and signal stability
  • No interference in biological samples, no pretreatments needed, and can be performed directly on raw biological samples
  • Store reagent and standard at 4°C and -20°C, respectively; shelf life of 12 months after receipt

Specifications

Content And Storage Store Reagent at 4°C. Store Standard at -20°C.
Description BCG Albumin Assay Kit is a quantitative colorimetric albumin determination at 620 nm.
Sample Type Plasma, Serum, Urine
Type BCG Albumin Assay Kit
For Use With (Application) Functional Study
Includes 50mL Reagent, Albumin standard, 1mL 5g/dL BSA
Quantity Each