missing translation for 'onlineSavingsMsg'
Learn More

Abietic Acid 80.0+%, TCI America™

Catalog No. a000125g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A000125G 25 g
A0001500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A000125G Supplier TCI America Supplier No. A000125G
Only null left

Chemical Identifiers

CAS 514-10-3
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD03423567
InChI Key RSWGJHLUYNHPMX-ONCXSQPRSA-N
Synonym abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
PubChem CID 10569
ChEBI CHEBI:28987
IUPAC Name (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
SMILES CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1

Specifications

Melting Point 164°C
Color White-Yellow
UN Number 3077
Formula Weight 302.46
Percent Purity ≥80.0% (GC)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Abietic Acid
EINECSNumber (4)-0675
RTECSNumber TP8580000
TSCA Yes