missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One ABI Fast Design System Compatible 96-well Microplates
Description
- Manufactured from medical grade polypropylene
- Half-skirt feature
Specifications
Specifications
| Array | 8 x 12 |
| Color | Natural |
| For Use With (Equipment) | ABI Fast Design System |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Quantity | 100/Cs. |