Learn More
Thermo Scientific™ Abgene™ SmartCap Mats
The Thermo Scientific™ Abgene™ SmartCap Mat ensures reliable sealing from ambient temperatures down to -80 °C.
Supplier:  Thermo Scientific™ AB1413
Catalog No. AB1413
Only null left
Explore available promotions
Description
Description
The Thermo Scientific™ Abgene™ SmartCap Mat seals 96 samples at one time in a high-density storage format,affording maximum space saving benefits. Suitable for use with Thermo Scientific storage tubes ranging from 0.3 ml to1.4 ml, the Abgene SmartCap Mat maintain sa consistent seal down to -80°C.
Features:
  • Efficient Storage
  • Enhanced Materials
  • Ease of Installation and Access
Specifications
Specifications
Seals 96 sample tubes at one time
96
Sealing Mat
-80°C
Product Suggestions
Product Suggestions