Learn More
Thermo Scientific™ Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Streamline your molecular biology applications with these 96-well Polypropylene Storage Plates.
Supplier: Thermo Scientific™ AB1058
Description
Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Thermo Scientific ABgene Storage and Assay Plates are manufactured in a Class 100,000 cleanroom environment to ensure that each plate is contamination-free. Plates are made using virgin medical grade polypropylene and have highly polished surfaces, making them low binding and chemically resistant. Rigid side walls and industry standard footprint make plates suitable for automation and robotic handling, as well as for low-throughput applications.
Highlights
- 96 Round wells with 200μL maximum well volume
- Reinforced skirt for robotic manipulation
- Seal with heat seals, adhesive seals, or PCR cap strips
- Raised rim design around each well for optimal sealing and reduced risk of cross-contamination
- Suitable for use with DMSO, Ethanol, and Isopropanol
- Alphanumeric grid-referencing
- Designed according to SBS footprint standards for robotic compatibility
- Wide working temperature range from -196° to +121°C
Specifications
| Custom Barcoding Available | |
| Standard | |
| 96-well Polypropylene Sample Processing & Storage Plates | |
| Autoclavable | |
| Low binding PP | |
| Natural | |
| ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
| Polypropylene | |
| 96 | |
| Cleanroom | |
| Round, Conical Bottom (V-well) | |
| Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
| 100 | |
| For Research Use Only. Not for use in diagnostic procedures. | |
| PCR Ready |
| Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
| Without Lid | |
| 8 x 12 | |
| Bulk | |
| Abgene | |
| 96-well plate | |
| Full | |
| 200 μL | |
| 200 μL | |
| Non-treated | |
| Yes | |
| 5000 x g | |
| 1 | |
| Yes | |
| ANSI/ SLAS Standard |