Learn More
Thermo Scientific™ Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Streamline your molecular biology applications with these 96-well Polypropylene Storage Plates.
Supplier: Thermo Scientific™ AB0796
Description
Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Highlights
- U-shaped well bottom
- Alphanumeric grid-referencing and industry standard footprint
- Highly polished, providing good well visibility
- Supporting ribs to aid stacking and improve centrifugation performance
Specifications
| Custom Barcoding Available | |
| Standard | |
| 8 x 12 | |
| Bulk | |
| Abgene | |
| 96-well plate | |
| Polypropylene | |
| 96 | |
| Cleanroom | |
| Round | |
| Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
| 100 | |
| For Research Use Only. Not for use in diagnostic procedures. | |
| PCR Ready |
| Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
| 96-well Polypropylene Sample Processing & Storage Plates | |
| Autoclavable | |
| Low binding PP | |
| Natural | |
| ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
| 330 μL | |
| 300 μL | |
| Non-treated | |
| Yes | |
| 5000 x g | |
| 1 | |
| Yes | |
| ANSI/ SLAS Standard |