Learn More
Thermo Scientific™ Abgene™ 96 Well 2.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
GREENER_CHOICE
Catalog No. AB0661
Change view
Click to view available options
Color:
Natural
Well Shape:
Square, Conical Bottom (V-well)
Square, Round Bottom (U-well)
Packaging:
Bulk
Individually Wrapped
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape Packaging
AB0661 Natural Square, Round Bottom (U-well) Individually Wrapped
AB0932 Natural Square, Conical Bottom (V-well) Individually Wrapped
AB0788 Natural Square, Round Bottom (U-well) Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AB0661 Supplier Thermo Scientific™ Supplier No. AB0661
Edge
2.2 mL deepwell storage plates for the culture or storage of large samples.
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Highlights
  • Designed for optimal sample recovery
  • Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
  • U-bottom (AB-0661) to aid resuspension or conical bottom (AB-0932) for ease of sample retrieval
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Description 96-well Polypropylene Sample Processing & Storage Plates
Array 8 x 12
Autoclavable Autoclavable
Packaging Individually Wrapped
Binding Type Low binding PP
Product Line Abgene
Color Natural
Product Type 96-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 2200 μL
No. of Wells 96
Volume (Metric) Working 1900 μL
Sterility Sterile
Surface Treatment Non-treated
Well Shape Square, Round Bottom (U-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Green Features ACT Ecolabel (Silver)
Max. RCF 5000 x g
No. per Case 50
No. per Pack 1
Quantity 50/Cs.
Regulatory Status RUO
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard
Show More Show Less