missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
GREENER_CHOICE

Catalog No. p-6061266
Change view
Click to view available options
Well Shape:
Round
Square, Round Bottom (U-well)
Packaging:
Bulk
Individually Wrapped
Fit for Purpose:
PCR Ready
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Packaging Fit for Purpose
AB1127 Square, Round Bottom (U-well) Individually Wrapped PCR Ready
AB0564 Round Individually Wrapped PCR Ready
AB0787 Round Bulk PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AB1127 Supplier Thermo Scientific™ Supplier No. AB1127
Only null left

Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

Highlights

  • Round well shape designed for optimal sample recovery or square shape to maximize sample volume within ANSI footprint design
  • Each well has an independent sealing rim to prevent cross-contamination
  • U-bottomed wells ideally suited for sample resuspension
  • Polypropylene (AB-1068, polystyrene)
  • Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
  • Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
  • Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
  • Cleanroom manufactured
  • ANSI-format for compatibility with automated systems

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Description 96-well Polypropylene Sample Processing & Storage Plates
Array 8 x 12
Autoclavable Autoclavable
Packaging Individually Wrapped
Binding Type Low binding PP
Product Line Abgene
Color Natural
Product Type 96-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 1200 μL
No. of Wells 96
Volume (Metric) Working 1000 μL
Sterility Cleanroom
Surface Treatment Non-treated
Well Shape Square, Round Bottom (U-well)
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF 2000 x g
No. per Case 50
No. per Pack 1
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard
Show More Show Less