missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo-Fast PCR Plate, 48-well, clear

Catalog No. AB0648
Change view
Click to view available options
Quantity:
50 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0648 50 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0648 Supplier Thermo Scientific™ Supplier No. AB0648

Optimize PCR and qPCR applications with 48-well semi-skirted plates.

48-well semi-skirted plate for PCR and qPCR applications comes in a convenient format, which allows for lower sample numbers. The semi skirt makes the plate more rigid and allots space for labeling and barcoding. Available in assorted colors, it's compatible with standard multichannel pipettes and automated liquid handling systems.The plate is sealable when used with adhesive films, foils or cap strips.

Highlights

  • Convenient format for lower sample numbers
  • Semi-skirt adds rigidity and allows for labeling or barcoding
  • Sealable using adhesive films and foils and flat, ultra clear and domed cap strips
  • 0.3 mL maximum well volume (when used with adhesive and heat seals)

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 50 Plate
Color Clear
Product Type PCR Microplate
For Use With (Equipment) Thermal Cycler
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 0.3 mL
No. of Wells 48
Volume (Metric) Working 0.2 mL
Quantity 50 plates
Reaction Speed Standard
Show More Show Less