missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 384 Well Polypropylene Sample Processing & Storage Plates
GREENER_CHOICE

384-well microarray plate with pyramidal well bottoms for maximum sample recovery.

$1,241.32 - $2,346.11

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Description 384-well Polypropylene Sample Processing & Storage Plates
Array 16 x 24
View More Specs

Products 4
Catalog Number Mfr. No. Color Volume (Metric) Well Well Shape Fit for Purpose Price Quantity & Availability  
Catalog Number Mfr. No. Color Volume (Metric) Well Well Shape Fit for Purpose Price Quantity & Availability  
AB0781
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
AB0781
Natural 120 μL Square, Conical Bottom (V-well) PCR Ready
Case of 50 Each for $2,003.93
Only null left
 
AB1178
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
AB1178
Natural 250 μL Square, Conical Bottom (V-well) PCR Ready
Case of 50 Each for $1,241.32
Only null left
 
AB0862
View Documents Promotion Details
Thermo Scientific™
AB0862
Natural 300 μL Square, Conical Bottom (V-well) PCR Ready
Case of 50 Each for $2,346.11
Only null left
 
AB1056
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
AB1056
Natural 55 μL Round, Conical Bottom (V-well) PCR Ready
Case of 50 Each for $1,568.18
Only null left
 
Description

Description

Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using USP <661.1> high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and DeepWell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

All Thermo Scientific plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

 

Highlights

  • Certified free from DNAse, RNAse, PCR Inhibitors, and human DNA
  • Pyramidal well bottom for ease of sample retrieval
  • Discrete raised rims around each well for effective sealing and to reduce the risk of cross-contamination

Compatible Sealing Film: Adhesive and Heat
Hazardous: No
Max Centrifugal Force: 2000-5000 RCF
Storage Condition: Ambient stored, or transported

All plates are ANSI format for compatibility with automated systems.

To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.

Specifications

Specifications

Custom Barcoding Available
Standard
16 x 24
Low binding PP
384-well plate
Polypropylene
Cleanroom
Yes
5000 x g
For Research Use Only. Not for use in diagnostic procedures.
PCR Ready
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
384-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Abgene
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
384
Non-treated
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
50
Yes
ANSI/ SLAS Standard
Videos
SDS
Documents

Documents