missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thermo-Fast PCR Plate, 24-well

Catalog No. AB0624
Change view
Click to view available options
Quantity:
50 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB0624 50 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB0624 Supplier Thermo Scientific™ Supplier No. AB0624
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Increase stability and ease of handling with Thermo Scientific™ ABgene™ 24-Well PCR Plates.

Thermo Scientific™ ABgene 24-Well PCR Plates are easier to use than multiple 8- or 12-well strips, as the plate format allows wells to be sealed with adhesive film or strip caps and also increases stability and ease of handling.

  • Conveniently pre-cut into 24-well segments
  • Semi-skirt adds rigidity and allows for labeling or bar coding
  • Thin wall polypropylene

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 50 Plate
Color Clear
Product Type PCR Microplate
For Use With (Equipment) Thermal Cycler
Skirt Style Semi
Material Polypropylene
No. of Wells 24
Volume (Metric) Working 0.2 mL
Quantity 50 plates
Reaction Speed Standard
Show More Show Less