missing translation for 'onlineSavingsMsg'
Learn More

ABD-F 98.0+%, TCI America™

Catalog No. p-7118290
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5597100MG 100 mg
A5597500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5597100MG Supplier TCI America Supplier No. A5597100MG
Only null left

[HPLC Labeling Reagent for Determination of Thiols]

Chemical Identifiers

CAS 91366-65-3
Molecular Formula C6H4FN3O3S
Molecular Weight (g/mol) 217.17
MDL Number MFCD00036687
InChI Key XROXHZMRDABMHS-UHFFFAOYSA-N
Synonym abd-f, 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzofurazan-4-sulfonamide, 4-fluoro-7-sulfamoylbenzofurazan, 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonamide, 4-aminosulphonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonamide, pubchem23230, 4-fluoro-7-sulfamoylbenzofurazane, 4-fluoro-7-sulfoamoylbenzofurazan
PubChem CID 122067
IUPAC Name 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(F)C2=NON=C12

Specifications

Melting Point 146°C
Color Yellow
Quantity 100 mg
Formula Weight 217.17
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] [HPLC Labeling Reagent for Determina