missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AbC™ Total Antibody Compensation Bead Kit

Provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated mouse, rat, hamster, or rabbit antibodies with any laser in flow cytometry.

Supplier:  Invitrogen™ A10497

Encompass

 View more versions of this product

Catalog No. A10497


Only null left

Description

Description

  • Broadest multispecies reactivity—for use with all isotypes of mouse, rat, hamster, and rabbit antibody conjugates
  • Brightest signal—most sensitive antibody-capture beads available
  • Maximum laser compatibility—for use with all lasers
  • Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads).
  • Ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3), rat (IgG1, IgG2a, IgG2b), hamster (Armenian IgG and Syrian IgG), and rabbit (monoclonal and polyclonal) immunoglobulins.Demonstrates the highest and most consistent reactivity to the different subclasses of immunoglobulin in the industry.
Specifications

Chemical Identifiers

6 μm
Tube
Fluorescence
Room Temperature
Compensation Beads
Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Flow Cytometer
    any lasers (UV to 633/635)
    AbC

    Specifications

    6 μm
    Tube
    100 Tests
    Fluorescence
    Room Temperature
    Compensation Beads
    Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Flow Cytometer
    100 tests
    any lasers (UV to 633/635)
    AbC
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions

    For Research Use Only. Not for use in diagnostic procedures.