missing translation for 'onlineSavingsMsg'
Learn More

Abacavir Impurity 1, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022861
Change view
Click to view available options
:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111022861 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022861 Supplier Merck Emd Millipore Supplier No. BP1114
Only null left

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 120503-69-7
Flash Point Not Applicable
Molecular Formula C8H10N6
Linear Formula C8H10N6
Quantity 20 mg
Synonym N6-Cyclopropyl-9 H-purine-2,6-diamine
Formula Weight 190.21
Grade British Pharmacopoeia (BP) Reference Standard