Learn More
Enzo Life Sciences alphaB Crystallin monoclonal antibody (1B6.1 3G4) (50 g) DFS Item
Supplier:  Enzo ADISPA222D
Clone: 1B6.1-3G4. Host: Mouse. Isotype: IgG1. Immunogen: Bovine B-crystallin. UniProt: P02510. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Chicken, Bovine. Applications: ICC, IF, IHC (PS), WB. Recommended dilutions: Immunohistochemistry (paraffin sections, 10g/ml). Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. NC1607417
Only null left
Explore available promotions